Merck
  • Effect of water storage and heat treatment on the cytotoxicity of soft liners.
Effect of water storage and heat treatment on the cytotoxicity of soft liners.
Gerodontology (2011-02-03)
Lidia Yileng Tay, Daniel Rodrigo Herrera, Cristiane Campos Costa Quishida, Iracilda Zeppone Carlos, Janaina Habib Jorge
ABSTRACT
To evaluate the effect of water storage time on the cytotoxicity of soft liners. Sample discs of soft liners Dentusoft, Dentuflex, Trusoft, Ufi-Gel-P and denture base acrylic resin Lucitone-550 were prepared and divided into four groups: GN: No treatment, G24: Stored in water at 37°C for 24 h; G48: Stored in water at 37°C for 48 h, GHW: Immersed in water at 55°C for 10 min. To analyse the cytotoxic effect, three samples of each group were placed in tubes with Dubelcco's Modified Eagle Mediums and incubated at 37°C for 24 h. During this period, the toxic substances were leached to the culture medium. The cytotoxicity was analysed quantitatively by the incorporation of radioactivity (3)H-thymidine checking the number of viable cells (synthesis of DNA). The data were statistically analysed using two-way anova and Tukey's honestly significant difference tests (α = 0.05). Treatments did not reduce the cytotoxicity effect of the soft liners (p > 0.05). It was found that Ufi-Gel-P had a non-cytotoxic effect, Trusoft had a slightly cytotoxic effect, Dentuflex had a moderated cytotoxic effect, Dentusoft alternated between slightly and non-cytotoxic effect, and Lucitone-550 had non-cytotoxic effect when stored in water for 48 h. The effect of water storage and the heat treatment did not reduce the cytotoxicity of the soft liners.