- Modulation of reparative regeneration and CD117 expression by liver cells after partial hepatectomy in mice.
Modulation of reparative regeneration and CD117 expression by liver cells after partial hepatectomy in mice.
Bulletin of experimental biology and medicine (2011-01-18)
B G Yushkov, I G Danilova, Zh B Ponezheva, I A Brykina, M T Abidov, O V Kalyuzhin
PMID21240352
ABSTRACT
The effects of 3-aminophthalhydrazide and carrageenan on reparative regeneration and expression of CD117 by liver cells after partial hepatectomy were studied in mice. 3-Aminophthalhydrazide stimulated regeneration of the liver and increased the count of CD117(+) hepatocytes. By contrast, carrageenan inhibited liver reparation and CD117 expression.