Merck
  • Reduced expression of β2 integrin genes in rat peripheral leukocytes by inhibiting postprandial hyperglycemia.
Reduced expression of β2 integrin genes in rat peripheral leukocytes by inhibiting postprandial hyperglycemia.
Bioscience, biotechnology, and biochemistry (2010-12-15)
Kazuki Mochizuki, Masaya Shimada, Yutaro Tanaka, Nanae Fukaya, Toshinao Goda
ABSTRACT
β(2) integrins (CD11s/CD18) promote the attachment of leukocytes to vascular endothelial cells. We performed in this study sucrose loading to rats with moderate postprandial hyperglycemia with/without once-daily dosing of the α-glucosidase inhibitor, miglitol, for 4 days under 4-h fasting conditions. The streptozotocin (STZ)-treated rats showed moderate postprandial hyperglycemia on days 1 and 4. The gene expression was higher for CD11a with fasting and 3-h postprandial on day 1, and with fasting on day 4, for CD11b with fasting on day 1 and 3-h postprandial on day 4, and for CD18 with fasting on days 1 and 4 in peripheral leukocytes from the STZ-treated rats than in peripheral leukocytes from the saline-treated rats. Miglitol reduced postprandial hyperglycemia and the gene expression of CD11a with fasting and of CD11b 3-h postprandial on day 4. These results indicate that inhibiting postprandial hyperglycemia reduced the mRNA expression of β(2) integrins in peripheral leukocytes of moderately postprandial hyperglycemic rats.