Merck
  • Design and synthesis of novel thiophenecarbohydrazide, thienopyrazole and thienopyrimidine derivatives as antioxidant and antitumor agents.
Design and synthesis of novel thiophenecarbohydrazide, thienopyrazole and thienopyrimidine derivatives as antioxidant and antitumor agents.
Acta pharmaceutica (Zagreb, Croatia) (2010-12-08)
Ameen Ali Abu-Hashem, Mohamed Fathy El-Shehry, Farid Abd-Elraheem Badria
ABSTRACT
2-Amino-5-acetyl-4-methyl-thiophene-3-carboxylic acid ethyl ester (1) and 5-acetyl-2-amino-4-methylthiophene-3-carbohydrazide (2) were synthesized and used as starting materials for the synthesis of new series of 1-(5-amino-4-(3,5-dimethyl-1H-pyrazole-1-carbonyl)-3-methylthiophen-2-yl) ethanone (3a), 1-(5-amino-4-(4-chloro-3,5-dimethyl-1H-pyrazole-1-carbonyl)-3-methylthiophen-2-yl) ethanone (3b), 1-(4-methyl-2-amino-5-acetylthiophene-3-carbonyl)pyrazolidine-3,5-dione (4), (Z)-N'-(4-methyl-2-amino-5-acetylthiophene-3-carbonyl) formohydrazonic acid (5a), (Z)-ethyl-N'-4-methyl-2-amino-5-acetylthiophene-3-carbonylformo hydrazonate (5b), 6-acetyl-3-amino-2,5-dimethylthieno[2,3-d]pyrimidin-4(3H)-one (8), 5-methyl-3-amino-2-mercapto-6-acetylthieno [2,3-d]pyrimidin-4(3H)-one (10) and 5-methyl-6-acetyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one (12) as potential antioxidant and antitumor agents. Pharmacological tests showed that compounds 6a, 6b, 8, 10 and 12 exhibited significant antitumor and antioxidant activity.