- Nearly monodispersed core-shell structural Fe3O4@DFUR-LDH submicro particles for magnetically controlled drug delivery and release.
Nearly monodispersed core-shell structural Fe3O4@DFUR-LDH submicro particles for magnetically controlled drug delivery and release.
Chemical communications (Cambridge, England) (2010-11-17)
Dengke Pan, Hui Zhang, Ting Fan, Jiangang Chen, Xue Duan
PMID21079851
ABSTRACT
Nearly monodispersed magnetic Fe(3)O(4)@DFUR-LDH submicro particles containing the anticancer agent DFUR were prepared via a coprecipitation-calcination-reconstruction strategy of LDH materials over the surface of Fe(3)O(4) particles, and present well-defined core-shell structure, strong magnetization and obvious magnetically controlled drug delivery and release properties.