Merck
  • Dermal uptake and excretion of 14C-toluene diisocyante (TDI) and 14C-methylene diphenyl diisocyanate (MDI) in male rats. Clinical signs and histopathology following dermal exposure of male rats to TDI.
Dermal uptake and excretion of 14C-toluene diisocyante (TDI) and 14C-methylene diphenyl diisocyanate (MDI) in male rats. Clinical signs and histopathology following dermal exposure of male rats to TDI.
Toxicology letters (2010-10-12)
H D Hoffmann, E Leibold, C Ehnes, E Fabian, R Landsiedel, A Gamer, A Poole
ABSTRACT
Polyurethanes (PU) are polymers made with diisocyanates such as MDI (4,4'-methylene diphenyl diisocyanate) and TDI (2,4-toluene diisocyanate and 2,6-toluene diisocyanate). Investigations have been undertaken with MDI and TDI to assess dermal uptake and resulting systemic exposure. Absorption, distribution and excretion of MDI was studied in rats using a single dermal administration of (14)C-MDI dissolved in acetone at nominal 165 mg/kg body weight and 15 mg/kg bw (4.0 and 0.4 mg/cm(2)) and intradermal injection of (14)C-MDI dissolved in corn oil at nominal 1.4 mg/kg bw. Dermal absorption of (14)C-MDI (at both doses) was low; at or below 1% of the applied dose. Considerable amounts of the applied radioactivity were found at the application site which could not be washed off. By intradermal administration of (14)C-MDI approximately 66% of applied radioactivity remained at the application site with approximately 26% recovered in excreta, cage wash, tissues and carcass. The absorption, distribution and excretion of 2,4-TDI was studied in rats following a single dermal administration of radiolabelled (14)C-2,4-TDI at nominal 350 mg/kg body weight (12 mg/cm(2)). Dermal absorption of (14)C-2,4-TDI was at or below 1% of the applied dose. Considerable amounts of the applied radioactivity were found at the application site which could not be washed off. In summary the results show that dermal uptake of MDI and TDI is very low. Due to the chemical reactivity of isocyanates it can be expected that small amounts which might be absorbed will react with tissue constituents directly at the exposed skin area, or will be converted to adducts with biomacromolecules or to biologically inactive oligoureas. Overall it is concluded that, following dermal exposure to MDI and TDI, systemic exposures and resulting toxicity, other than the known sensitization, can be expected to be very low. In addition studies were performed with dermal application of unlabelled 2,4 and 2,6 TDI to check the availability and fate of this chemical on rat skin surface and to assess possible tissue damage. These experiments showed that unchanged test material can be detected on rat skin for up to 8h if not washed off. Dermal treatment with 2,4 or 2,6 TDI was associated with irritation with increased severity over a 48 h period after washing with a decontaminant solution.
Sigma-Aldrich
4,4′-Methylenebis(phenyl isocyanate), 98%