Merck
  • A randomized, double-blind study comparing the efficacy of selenium sulfide shampoo 1% and ciclopirox shampoo 1% as adjunctive treatments for tinea capitis in children.
A randomized, double-blind study comparing the efficacy of selenium sulfide shampoo 1% and ciclopirox shampoo 1% as adjunctive treatments for tinea capitis in children.
Pediatric dermatology (2010-08-26)
Catherine Chen, Laine H Koch, James E Dice, Kimberly K Dempsey, Alan B Moskowitz, Myra L Barnes-Eley, Thomas W Hubbard, Judith V Williams
ABSTRACT
Our objective was to compare the efficacy of selenium sulfide shampoo 1% and ciclopirox shampoo 1% as adjunctive treatments for tinea capitis in children. Forty children aged 1-11 years with clinically diagnosed tinea capitis were randomized to receive selenium sulfide shampoo 1% or ciclopirox shampoo 1% twice a week as adjuncts to an 8-week course of ultramicronized griseofulvin dosed at 10-12 mg/kg/day. At weeks 2, 4, and 8, subjects returned to the clinic for evaluation and scalp cultures. Subjects then returned for follow-up visits 4 weeks after completing treatment. Overall, by 8 weeks, 30 of 33 (90.9%) treated children demonstrated mycological cure. Selenium sulfide shampoo 1% and ciclopirox shampoo 1% were equally effective as adjunctive treatments for tinea capitis in children in our study.