Merck
  • N(G)-Acyl-argininamides as NPY Y(1) receptor antagonists: Influence of structurally diverse acyl substituents on stability and affinity.
N(G)-Acyl-argininamides as NPY Y(1) receptor antagonists: Influence of structurally diverse acyl substituents on stability and affinity.
Bioorganic & medicinal chemistry (2010-08-07)
Stefan Weiss, Max Keller, Günther Bernhardt, Armin Buschauer, Burkhard König
ABSTRACT
N(G)-Acylated argininamides, covering a broad range of lipophilicity (calculated logD values: -1.8-12.5), were synthesized and investigated for NPY Y(1) receptor (Y(1)R) antagonism, Y(1)R affinity and stability in buffer (N(G)-deacylation, yielding BIBP 3226). Broad structural variation of substituents was tolerated. The K(i) (binding) and K(b) values (Y(1)R antagonism) varied from low nM to one-digit muM. Most of the compounds proved to be sufficiently stable at pH 7.4 over 90min to determine reliable pharmacological data in vitro. Exceptionally high instability was detected when a succinyl moiety was attached to the guanidine, probably, due to an intramolecular cleavage mechanism.