Merck
  • Adsorption of pyrantel pamoate on mercury from aqueous solutions: studies by stripping voltammetry.
Adsorption of pyrantel pamoate on mercury from aqueous solutions: studies by stripping voltammetry.
Journal of colloid and interface science (2010-07-21)
Vinod K Gupta, Rajeev Jain, N Jadon, K Radhapyari
ABSTRACT
Adsorption and electrochemical reduction of pyrantel pamoate are studied in Britton Robinson buffer medium at hanging mercury drop electrode (HMDE) by Adsorptive Stripping Voltammetric technique. The peak current shows a linear dependence with the drug concentration over the range 250 ng mL(-1) to 64 microg mL(-1). Applicability to assay the drug in urine samples is illustrated in the concentration range 5-20 microg mL(-1).
Sigma-Aldrich
Pyrantel pamoate, BioXtra