Merck
  • Fluorescent and electrochemical sensing of polyphosphate nucleotides by ferrocene functionalised with two Zn(II)(TACN)(pyrene) complexes.
Fluorescent and electrochemical sensing of polyphosphate nucleotides by ferrocene functionalised with two Zn(II)(TACN)(pyrene) complexes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-07-14)
Zhanghua Zeng, Angel A J Torriero, Alan M Bond, Leone Spiccia
ABSTRACT
The [Fc-bis{Zn(II)(TACN)(Py)}] complex, comprising two Zn(II)(TACN) ligands (Fc=ferrocene; Py=pyrene; TACN=1,4,7-triazacyclononane) bearing fluorescent pyrene chromophores linked by an electrochemically active ferrocene molecule has been synthesised in high yield through a multistep procedure. In the absence of the polyphosphate guest molecules, very weak excimer emission was observed, indicating that the two pyrene-bearing Zn(II)(TACN) units are arranged in a trans-like configuration with respect to the ferrocene bridging unit. Binding of a variety of polyphosphate anionic guests (PPi and nucleotides di- and triphosphate) promotes the interaction between pyrene units and results in an enhancement in excimer emission. Investigations of phosphate binding by (31)P NMR spectroscopy, fluorescence and electrochemical techniques confirmed a 1:1 stoichiometry for the binding of PPi and nucleotide polyphosphate anions to the bis(Zn(II)(TACN)) moiety of [Fc-bis{Zn(II)(TACN)(Py)}] and indicated that binding induces a trans to cis configuration rearrangement of the bis(Zn(II)(TACN)) complexes that is responsible for the enhancement of the pyrene excimer emission. Pyrophosphate was concluded to have the strongest affinity to [Fc-bis{Zn(II)(TACN)(Py)}] among the anions tested based on a six-fold fluorescence enhancement and 0.1 V negative shift in the potential of the ferrocene/ferrocenium couple. The binding constant for a variety of polyphosphate anions was determined from the change in the intensity of pyrene excimer emission with polyphosphate concentration, measured at 475 nm in CH(3)CN/Tris-HCl (1:9) buffer solution (10.0 mM, pH 7.4). These measurements confirmed that pyrophosphate binds more strongly (K(b)=(4.45+/-0.41) x 10(6) M(-1)) than the other nucleotide di- and triphosphates (K(b)=1-50 x 10(5) M(-1)) tested.