Merck
  • Comparative analysis of SOX3 protein orthologs: Expansion of homopolymeric amino acid tracts during vertebrate evolution.
Comparative analysis of SOX3 protein orthologs: Expansion of homopolymeric amino acid tracts during vertebrate evolution.
Biochemical genetics (2010-05-25)
Marija Mojsin, Natasa Kovacevic-Grujicic, Aleksandar Krstic, Jelena Popovic, Milena Milivojevic, Milena Stevanovic
ABSTRACT
To understand more fully the structure and evolution of the SOX3 protein, we comparatively analyzed its orthologs in vertebrates. Since complex disorders are associated with human SOX3 polyalanine expansions, our investigation focused on both compositional and evolutionary analysis of various homopolymeric amino acid tracts observed in SOX3 orthologs. Our analysis revealed that the observed homopolymeric alanine, glycine, and proline tracts are mammal-specific, except for one polyglycine tract present in birds. Since it is likely that the SOX3 protein acquired additional roles in brain development in Eutheria, we might speculate that development of novel brain functions during the course of evolution was affected, at least in part, by such structural-functional changes in the SOX3 protein.