Merck
  • Triazophos up-regulated gene expression in the female brown planthopper, Nilaparvata lugens.
Triazophos up-regulated gene expression in the female brown planthopper, Nilaparvata lugens.
Journal of insect physiology (2010-03-13)
Yan-Yuan Bao, Bao-Ling Li, Zhao-Bu Liu, Jian Xue, Zeng-Rong Zhu, Jia-An Cheng, Chuan-Xi Zhang
ABSTRACT
The widespread use of insecticides has caused the resurgence of the brown planthopper, Nilaparvata lugens, in Asia. In this study, we investigated an organo-phosphorous insecticide, triazophos, and its ability to induce gene expression variation in female N. lugens nymphs just before emergence. By using the suppression subtractive hybridization method, a triazophos-induced cDNA library was constructed. In total, 402 differentially expressed cDNA clones were obtained. Real-time qPCR analysis confirmed that triazophos up-regulated the expression of six candidate genes at the transcript level in nymphs on day 3 of the 5th instar. These genes encode N. lugens vitellogenin, bystin, multidrug resistance protein (MRP), purine nucleoside phosphorylase (PNP), pyrroline-5-carboxylate reductase (P5CR) and carboxylesterase. Our results imply that the up-regulation of these genes may be involved in the induction of N. lugens female reproduction or resistance to insecticides.
Product Description
Supelco
Triazophos, PESTANAL®, analytical standard