Merck
  • Structure of sodiated octa-glycine: IRMPD spectroscopy and molecular modeling.
Structure of sodiated octa-glycine: IRMPD spectroscopy and molecular modeling.
Journal of the American Society for Mass Spectrometry (2010-03-02)
David Semrouni, O Petru Balaj, Florent Calvo, Catarina F Correia, Carine Clavaguéra, Gilles Ohanessian
ABSTRACT
The structure of the sodiated peptide GGGGGGGG-Na(+) or G(8)-Na(+) was investigated by infrared multiple photon dissociation (IRMPD) spectroscopy and a combination of theoretical methods. IRMPD was carried out in both the fingerprint and N-H/O-H stretching regions. Modeling used the polarizable force field AMOEBA in conjunction with the replica-exchange molecular dynamics (REMD) method, allowing an efficient exploration of the potential energy surface. Geometries and energetics were further refined at B3LYP-D and MP2 quantum chemical levels. The IRMPD spectra indicate that there is no free C-terminus OH and that several N-Hs are free of hydrogen bonding, while several others are bound, however not very strongly. The structure must then be either of the charge solvation (CS) type with a hydrogen-bound acidic OH, or a salt bridge (SB). Extensive REMD searches generated several low-energy structures of both types. The most stable structures of each type are computed to be very close in energy. The computed energy barrier separating these structures is small enough that G(8)-Na(+) is likely fluxional with easy proton transfer between the two peptide termini. There is, however, good agreement between experiment and computations in the entire spectral range for the CS isomer only, which thus appears to be the most likely structure of G(8)-Na(+) at room temperature.