Merck
Fragrance material review on 2-methyl-3-buten-2-ol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-02-10)
D McGinty, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of 2-methyl-3-buten-2-ol when used as a fragrance ingredient is presented.