Merck
  • Aporphine alkaloids, clerodane diterpenes, and other constituents from Tinospora cordifolia.
Aporphine alkaloids, clerodane diterpenes, and other constituents from Tinospora cordifolia.
Fitoterapia (2010-01-19)
Van Kiem Phan, Van Minh Chau, Tien Dat Nguyen, Van Kinh La, Thuy Hang Dan, Hoai Nam Nguyen, Xuan Cuong Nguyen, Thanh Huong Hoang, Van Lau Trinh
ABSTRACT
Phytochemical investigation of the methanol extract of Tinospora cordifolia aerial parts led to the isolation of four new and seven known compounds. The structures of two new aporphine alkaloids, N-formylasimilobine 2-O-beta-D-glucopyranosyl-(1-->2)-beta-D-glucopyranoside (tinoscorside A, 1) and N-acetylasimilobine 2-O-beta-D-glucopyranosyl-(1-->2)-beta-D-glucopyranoside (tinoscorside B, 2), a new clerodane diterpene, tinoscorside C (3), and a new phenylpropanoid, sinapyl 4-O-beta-D-apiofuranosyl-(1-->6)-O-beta-D-glucopyranoside (tinoscorside D, 6) were determined by extensive spectroscopic methods including FTICR-MS and 1D and 2D NMR.