Merck
  • Practical asymmetric conjugate alkynylation of Meldrum's acid-derived acceptors: access to chiral beta-alkynyl acids.
Practical asymmetric conjugate alkynylation of Meldrum's acid-derived acceptors: access to chiral beta-alkynyl acids.
Journal of the American Chemical Society (2009-12-22)
Sheng Cui, Shawn D Walker, Jacqueline C S Woo, Christopher J Borths, Herschel Mukherjee, Maosheng J Chen, Margaret M Faul
ABSTRACT
The enantioselective conjugate addition of alkynyl nucleophiles has been a long-standing challenge in synthetic chemistry. This paper describes a highly practical asymmetric conjugate alkynylation of Meldrum's acid-derived acceptors using cinchonidine (<$100/kg) as the chiral mediator. The process provides practical access to chiral beta-alkynyl acids. Noteworthy attributes of the method are its broad scope, high functional-group compatibility, and ease of scalability.