Merck
  • Temperature-dependent interfacial properties of hydrophobically end-modified poly(2-isopropyl-2-oxazoline)s assemblies at the air/water interface and on solid substrates.
Temperature-dependent interfacial properties of hydrophobically end-modified poly(2-isopropyl-2-oxazoline)s assemblies at the air/water interface and on solid substrates.
Journal of colloid and interface science (2009-10-03)
Rodolphe Obeid, Jin-Young Park, Rigoberto C Advincula, Françoise M Winnik
ABSTRACT
We describe herein the properties at the air/water (A/W) interface of hydrophobically end-modified (HM) poly(2-isopropyl-2-oxazoline)s (PiPrOx) bearing an n-octadecyl chain on both termini (telechelic HM-PiPrOx) or on one chain end (semitelechelic HM-PiPrOx) for different subphase temperatures and spreading solvents using the Langmuir film balance technique. The polymer interfacial properties revealed by the pi-A isotherms depend markedly on the architecture and molecular weight of the polymer. On cold water subphases (14 degrees C), diffusion of PiPrOx chains onto water takes place for all polymers in the intermediate compressibility region (5mNm(-1)). At higher subphase temperatures (36 and 48 degrees C), the HM-PiPrOx film exhibited remarkable stability with time. Brewster angle microscopy (BAM) imaging of the A/W interface showed that the polymer assembly was not uniform and that large domains formed, either isolated grains or pearl necklaces, depending on the polymer structure, the concentration of the spreading solution and the subphase temperature. The Langmuir films were transferred onto hydrophilic substrates (silica) by the Langmuir-Blodgett (LB) technique and onto hydrophobic substrates (gold) by Langmuir-Schaefer (LS) film deposition, resulting in the formation of adsorbed particles ranging in size from 200 to 500nm, depending on the polymer architecture and the substrate temperature. The particles presented "Janus"-like hydrophilic/hydrophobic characteristics.
Sigma-Aldrich
1-Butanethiol, ≥98%