Merck
  • [Misuse of substances theoretically without abuse potential--case series].
[Misuse of substances theoretically without abuse potential--case series].
Przeglad lekarski (2009-10-01)
Jacek Sein Anand, Bogusław Habrat, Michał Sut, Roman Korolkiewicz
ABSTRACT
The aim of our study was to present cases of misuse of different substances theoretically without abuse potential. In the last few years such behavior became an increasing problem in toxicological and emergency units. Lack of typical signs of intoxication with psychoactive substances, and negative results of standard toxicological tests may be a challenge for toxicologists and emergency medicine practitioners.