Merck
  • N-vanillylnonanamide tested as a non-toxic antifoulant, applied to surfaces in a polyurethane coating.
N-vanillylnonanamide tested as a non-toxic antifoulant, applied to surfaces in a polyurethane coating.
Biotechnology letters (2009-06-03)
Federica Villa, Lucia Giacomucci, Andrea Polo, Pamela Principi, Lucia Toniolo, Marienella Levi, Stefano Turri, Francesca Cappitelli
ABSTRACT
The potential on N-vanillylnonanamide (NVN) in preventing the attachment of Pseudomonas stutzeri and a Bacillus cereus-group strain was investigated. NVN up to 852 microM was not toxic, nor was it an energy source for either organism. Microbial attachment assays were carried out on glass and polylysine slides. with NVN being dispersed in or applied to the surfaces using a polyurethane coating. NVN at 205 microM inhibited Bacillus adhesion on glass slides by 48% and the percentage did not significantly increase at 852 microM. NVN blended into or sprayed onto the coating at 205 micromol/kg did not prevent adhesion. The compound is therefore not useful as an antifouling product under the tested coating conditions.