Merck
  • A general method for the synthesis of 3,5-diarylcyclopentenones via friedel-crafts acylation of vinyl chlorides.
A general method for the synthesis of 3,5-diarylcyclopentenones via friedel-crafts acylation of vinyl chlorides.
The Journal of organic chemistry (2009-06-03)
Yingju Xu, Mark McLaughlin, Cheng-yi Chen, Robert A Reamer, Peter G Dormer, Ian W Davies
ABSTRACT
A general approach for the synthesis of 3,5-diarylcyclopentenones was developed. Key aspects of this approach are the intramolecular Friedel-Crafts-type cyclization of vinyl chlorides and subsequent Pd-catalyzed cross-coupling reactions. The requisite vinyl chloride-bearing arylacetic acid precursors are readily available by straightforward alkylation of arylacetic acid esters and undergo cyclization to yield 3-chloro-5-aryl-2-cyclopentenones when treated with AlCl(3). The vinylogous acid chloride functionality present in these immediate products allows for further elaboration via Pd-catalyzed cross-coupling chemistry, leading to a diverse array of products.
Sigma-Aldrich
Ethyl vinyl ketone, ≥97%, stabilized with BHT, FG