Merck
Species difference of site-selective glucuronidation of morphine.
Journal of pharmacobio-dynamics (1991-04-01)
C K Kuo, N Hanioka, Y Hoshikawa, K Oguri, H Yoshimura
ABSTRACT
Species difference in glucuronidation of morphine was studied using mice, rats, guinea pigs and rabbits in vivo and in vitro. Morphine-3-glucuronide (M-3-G) and morphine-6-glucuronide (M-6-G) were determined by high-performance liquid chromatography. M-3-G was the major urinary metabolite of morphine in all these animal species. However, a remarkable species difference was observed in the urinary excretion of the M-6-G. Excretion ratios of the M-3-G to M-6-G were approximately 4:1 and 50:1 in guinea pigs and rabbits, respectively. The urinary excretion of M-6-G in mice and rats was too small to be determined. On the other hand, the ratios of uridine diphosphate-glucuronyltransferase (UDPGT) activities toward 3- and 6-hydroxyl groups of morphine in liver microsomes of mice, rats, guinea pigs and rabbits were approximately 300:1, 90:1, 4:1 and 40:1, respectively. Ratios of two morphine UDPGT activities in the liver microsomes of guinea pigs and rabbits, thus, reflected those of urinary excretion of morphine glucuronides.
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%