Merck
  • Paget's disease 2: exploring diagnosis, management and support strategies.
Paget's disease 2: exploring diagnosis, management and support strategies.
Nursing times (2009-03-31)
Anne Sutcliffe
ABSTRACT
This is the second of a two-part unit on Paget's disease. Part 1 outlined the epidemiology, pathophysiology, causes and clinical features of the condition. This part outlines advice on diagnosis, drug therapy, surgery, other management options and emotional support.