- Computational evaluation of enantioselective Diels-Alder reactions mediated by Corey's cationic oxazaborolidine catalysts.
Computational evaluation of enantioselective Diels-Alder reactions mediated by Corey's cationic oxazaborolidine catalysts.
The Journal of organic chemistry (2008-12-11)
Michael N Paddon-Row, Christopher D Anderson, K N Houk
PMID19067584
ABSTRACT
The cationic oxazaborolidine-catalyzed Diels-Alder reactions of butadiene and a series of five dienophiles have been studied using density functional theory (B3LYP/6-31G(d)). In each case, the computational results successfully reproduce the experimentally observed sense of stereoinduction and enantiomeric ratio. The computed structures of the lowest energy Lewis acid-carbonyl complexes and Lewis acid-transition state complexes are closely related to the models for stereoselection proposed by Corey and co-workers.