Merck
  • FBX aqueous chemical dosimeter for measurement of virtual wedge profiles.
FBX aqueous chemical dosimeter for measurement of virtual wedge profiles.
Journal of applied clinical medical physics (2008-11-21)
Manoj K Semwal, Anil K Bansal, Pradeep K Thakur, Pandit B Vidyasagar
ABSTRACT
We investigated the ferrous sulfate-benzoic acid-xylenol orange (FBX) aqueous chemical dosimeter for measurement of virtual (dynamic) wedge profiles on a linear accelerator. The layout for irradiation of the FBX-filled tubes mimicked a conventional linear detector array geometry. A comparison of the resulting measurements with film-measured profiles showed that, in the main beam region, the difference between the FBX system and the film system was within +/-2% and that, in the penumbra region, the difference varied from +/-1 mm to +/-2.5 mm in terms of positional equivalence, depending on the size of the dosimeter tubes. We thus believe that the energy-independent FBX dosimetry system can measure virtual wedge profiles with reasonable accuracy at reasonable cost. However, efficiency improvement is required before this dosimetry system can be accepted into routine practice.