- Segmental order parameters in a nonionic surfactant lamellar phase studied with 1H-13C solid-state NMR.
Segmental order parameters in a nonionic surfactant lamellar phase studied with 1H-13C solid-state NMR.
Physical chemistry chemical physics : PCCP (2008-10-01)
Tiago Mendes Ferreira, Bruno Medronho, Rachel W Martin, Daniel Topgaard
PMID18825291
ABSTRACT
A lyotropic nonionic lamellar system composed of pentaethyleneglycol mono n-dodecyl ether and D(2)O was studied using natural abundance (13)C NMR under magic-angle spinning. Applying a two-dimensional recoupling method proposed by Dvinskikh (R-PDLF), (1)H-(13)C dipolar couplings were estimated over a range of temperatures (300-335 K), thus enabling analysis of structural changes in the liquid crystalline system. The results obtained are used to correlate the conformation and mobility of local sites in the surfactant molecule with overall changes in the lamellar structure.