Merck
  • Comparative effects of herbicides on photosynthesis and growth of tropical estuarine microalgae.
Comparative effects of herbicides on photosynthesis and growth of tropical estuarine microalgae.
Marine pollution bulletin (2008-07-18)
Marie Magnusson, Kirsten Heimann, Andrew P Negri
ABSTRACT
Pulse amplitude modulation (PAM) fluorometry is ideally suited to measure the sub-lethal impacts of photosystem II (PSII)-inhibiting herbicides on microalgae, but key relationships between effective quantum yield [Y(II)] and the traditional endpoints growth rate (micro) and biomass increase are unknown. The effects of three PSII-inhibiting herbicides; diuron, hexazinone and atrazine, were examined on two tropical benthic microalgae; Navicula sp. (Heterokontophyta) and Nephroselmis pyriformis (Chlorophyta). The relationships between Y(II), micro and biomass increase were consistent (r2 > or =0.90) and linear (1:1), validating the utility of PAM fluorometry as a rapid and reliable technique to measure sub-lethal toxicity thresholds of PSII-inhibiting herbicides in these microalgae. The order of toxicity (EC50 range) was: diuron (16-33 nM) > hexazinone (25-110 nM) > atrazine (130-620 nm) for both algal species. Growth rate and photosynthesis were affected at diuron concentrations that have been detected in coastal areas of the Great Barrier Reef.