Merck
  • Effect of a phosphodiesterase type 3 inhibitor in oocyte maturation medium on subsequent mouse embryo development.
Effect of a phosphodiesterase type 3 inhibitor in oocyte maturation medium on subsequent mouse embryo development.
Fertility and sterility (2008-06-17)
Byung Chul Jee, Hai-Ying Chen, Ri-Cheng Chian
ABSTRACT
To investigate the effect of a phosphodiesterase type 3 inhibitor in oocyte maturation medium on subsequent embryo development. Animal study. University laboratory. CD-1 mice. Immature oocytes were arrested meiotically in medium containing a specific phosphodiesterase type 3 inhibitor, cilostamide (1 micromol/L, 5 micromol/L, and 10 micromol/L), for 6 and 24 hours of prematuration culture, and then matured in vitro in inhibitor-free medium. Oocyte maturation, fertilization, and early embryonic development in vitro. After 6 and 24 hours of prematuration culture with 1 micromol/L, 5 micromol/L, and 10 micromol/L cilostamide, there were no differences in maturation, fertilization, and early embryonic development compared with non-arrested controls. Prematuration culture with cilostamide, for either 6 or 24 hours, may not be beneficial for subsequent embryonic development of immature mouse oocytes.