Merck
  • Stepwise two-color laser photolysis studies of alpha-cleavage in highly excited triplet states of alpha-acyl-4-phenylphenols.
Stepwise two-color laser photolysis studies of alpha-cleavage in highly excited triplet states of alpha-acyl-4-phenylphenols.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2008-06-06)
Minoru Yamaji
ABSTRACT
The photochemical properties of alpha-cleavage of C-O bond in highly excited triplet states (T(n) with n>2) of p-biphenyl acetate and p-biphenyl benzoate (Me-OBP and Ph-OBP) in solution were investigated in comparison with those in the lowest excited singlet and triplet states by using single laser and sequential two-color two-laser photolysis techniques. Upon 266 nm laser photolysis of Me-OBP and Ph-OBP, occurrence of C-O bond cleavage in the excited singlet state was recognized from the observation of the formation of p-phenylphenoxy radical (PPR) in the transient absorption. The quantum yields (Phi(rad)) of the PPR formation were determined to be 0.29 and 0.24 for Me-OBP and Ph-OBP, respectively. Triplet sensitization using acetone (Ac) provided efficient formation of the lowest triplet states (T(1)) of Me-OBP and Ph-OBP, and the molar absorption coefficients of the triplet-triplet absorption were determined. No photochemical reactions were found in the T(1) state. Upon 355 nm laser flash photolysis of the T(1) states of Me-OBP and Ph-OBP, formation of PPR accompanied with decomposition of the triplet state was confirmed in the transient absorption. This observation indicated that alpha-cleavage proceeds in the highly excited triplet state. The quantum yields (Phi(dec)) of the decomposition in the dissociative highly excited triplet state (T(R)) were determined to be 0.25 and 0.15 for Me-OBP and Ph-OBP, respectively. The reaction mechanism for alpha-bond cleavage in the T(R) state was discussed.
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%