- A novel protecting/activating strategy for beta-hydroxy acids and its use in convergent peptide synthesis.
A novel protecting/activating strategy for beta-hydroxy acids and its use in convergent peptide synthesis.
The Journal of organic chemistry (2008-02-22)
Jan Spengler, Javier Ruíz-Rodríguez, Francesc Yraola, Miriam Royo, Manfred Winter, Klaus Burger, Fernando Albericio
PMID18288867
ABSTRACT
beta-Hydroxy acids were reacted with hexafluoroacetone and carbodiimides to give carboxy-activated six-membered lactones in good yields. On reaction with amines, the corresponding amides were obtained. We demonstrate the following applications of this protecting/activating strategy: preparation of carboxamides in solution and on solid phase (both normal and reverse mode); recovery and reuse of the excess material in solid-phase synthesis; and convergent solid-phase peptide synthesis (CSPPS) with peptide segments bearing C-terminal Ser or Thr with very low levels of epimerization (<1%, HPLC).