Merck
  • Headache caused by handling the pesticide Antracol Combi WP 76: a case report.
Headache caused by handling the pesticide Antracol Combi WP 76: a case report.
The journal of headache and pain (2007-12-13)
Marina Titlic, Ivana Jukic, Ante Tonkic, Ante Punda
ABSTRACT
We report on headache caused by the pesticide Antracol Combi WP 76. A headache caused by handling of pesticides should be distinguishable from migraine and headaches from other causes. This 29-year-old woman, who handled pesticides, presented with very strong headache with nausea and vomiting. Poisoning by this pesticide cannot be proven toxicologically, and there is no antidote. Following isolation from the pesticide and abundant hydration, the pain gradually decreased. History, clinical signs and neuroradiological examinations excluded other possible causes of the headache. In treatment of patients in rural areas, attention should also be paid to the pesticide Antracol Combi WP 76 as a possible cause of headaches.