Merck
  • Biochemical studies on the terrestrial snail, Eubania vermiculata (Müller) treated with some pesticides.
Biochemical studies on the terrestrial snail, Eubania vermiculata (Müller) treated with some pesticides.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (1991-01-01)
H B el-Wakil, M A Radwan
ABSTRACT
The in vivo effects of methomyl, thiodicarb and metaldehyde on total soluble proteins, total lipids and glycogen content, in addition, the activity of glutamic oxaloacetic transaminase, (GOT), (GPT) glutamic pyruvic transaminase and catalase (CAT) enzymes of terrestrial E. vermiculata snails was studied. The experimental snails were treated with low concentration of 0.2% brain bait w/w of the pesticides for a period of 1,3,5,7 and 10 days. The results showed that methomyl and thiodicarb lead to significant reduction in total soluble proteins, lipids, and glycogen content, while significant increases in the activity of all enzymes tested were noted. Metaldehyde treatment showed no significant effect on total soluble proteins, lipids and GOT level, whereas a significant increase in GPT and CAT enzymes was observed. Also, metaldehyde resulted a significant reduction in glycogen content of snails.