- Comparison of the volatile constituents in cold-pressed bergamot oil and a volatile oil isolated by vacuum distillation.
Comparison of the volatile constituents in cold-pressed bergamot oil and a volatile oil isolated by vacuum distillation.
Journal of agricultural and food chemistry (2007-08-19)
Emilia L Belsito, Concetta Carbone, Maria L Di Gioia, Antonella Leggio, Angelo Liguori, Francesca Perri, Carlo Siciliano, Maria C Viscomi
PMID17696485
ABSTRACT
The vacuum distillation of bergamot peels furnishes a high-quality essential oil that is totally bergapten-free. This oil was compared with that produced by distillation of cold-pressed oils and those commercially available. The oil obtained by vacuum distillation of the bergamot vegetable matrix shows a composition quite similar to that of the cold-pressed oil. It also displays qualitative characteristics that are superior with respect to those normally observed for essential oils isolated by distillation of cold-pressed oils. Oils isolated by the method presented here can constitute ideal candidates in producing foods, for example, Earl Grey tea, and cosmetic preparations.