Merck
  • Nifuroxazide-induced acute pancreatitis: a new side-effect for an old drug?
Nifuroxazide-induced acute pancreatitis: a new side-effect for an old drug?
Acta gastro-enterologica Belgica (2007-07-11)
Akilimali Shindano, Liliane Marot, André P Geubel
ABSTRACT
We report the case of a middle-aged woman who developed a typical picture of acute pancreatitis together with systemic features of immunoallergy after the intake of two capsules (200 mg) of nifuroxazide. Even if acute pancreatitis is a rare adverse event of nitrofuran derivative therapy, nifuroxazide-induced pancreatitis as not been previously described. As suggested by associated systemic features, the disease is likely of immunoallergic origin.