Merck
  • An attempt to design an isoluminol-hydrogen peroxidase-amplified CL that measures intracellularly produced H2O2 in phagocytes: sensitivity for H2O2 is not high enough to allow detection.
An attempt to design an isoluminol-hydrogen peroxidase-amplified CL that measures intracellularly produced H2O2 in phagocytes: sensitivity for H2O2 is not high enough to allow detection.
Luminescence : the journal of biological and chemical luminescence (2007-07-05)
Anna-Lena Stenfeldt, Claes Dahlgren
ABSTRACT
A technique was designed for the determination of hydrogen peroxide release. We found, however, that the isoluminol-amplified chemiluminescence technique is a suitable tool for measuring secretion of superoxide but not hydrogen peroxide.