- Highly efficient three-component synthesis of beta-lactams from N-methylhydroxylamine, aldehydes, and phenylacetylene.
Highly efficient three-component synthesis of beta-lactams from N-methylhydroxylamine, aldehydes, and phenylacetylene.
Chemistry, an Asian journal (2007-04-19)
Liang Zhao, Chao-Jun Li
PMID17441056
ABSTRACT
The three-component reaction of N-substituted hydroxylamines, aldehydes, and phenylacetylene catalyzed by CuCl/2,2'-bipyridine in the presence of NaOAc under neat conditions afforded the corresponding beta-lactams in good to excellent yields. Aromatic, heteroaromatic, and aliphatic aldehydes are tolerated in this reaction. The electronic effects of the aldehydes were studied for the reaction with N-methylhydroxylamine and N-benzylhydroxylamine. For N-methylhydroxylamine, electron-rich aldehydes provided higher yields than electron-deficient aldehydes, whereas for N-benzylhydroxylamine, no significant electronic effect was observed for the aldehydes.