Merck
  • Design, synthesis, and X-ray structure of potent memapsin 2 (beta-secretase) inhibitors with isophthalamide derivatives as the P2-P3-ligands.
Design, synthesis, and X-ray structure of potent memapsin 2 (beta-secretase) inhibitors with isophthalamide derivatives as the P2-P3-ligands.
Journal of medicinal chemistry (2007-04-17)
Arun K Ghosh, Nagaswamy Kumaragurubaran, Lin Hong, Sarang S Kulkarni, Xiaoming Xu, Wanpin Chang, Vajira Weerasena, Robert Turner, Gerald Koelsch, Geoffrey Bilcer, Jordan Tang
ABSTRACT
Structure-based design and synthesis of a number of potent and selective memapsin 2 inhibitors are described. These inhibitors were designed based upon the X-ray structure of memapsin 2-bound inhibitor 3 that incorporates methylsulfonyl alanine as the P2-ligand and a substituted pyrazole as the P3-ligand. Of particular importance, we examined the ability of the substituted isophthalic acid amide derivative to mimic the key interactions in the S2-S3 regions of the enzyme active sites of 3-bound memapsin 2. We investigated various substituted phenylethyl, alpha-methylbenzyl, and oxazolylmethyl groups as the P3-ligands. A number of inhibitors exhibited very potent inhibitory activity against mempasin 2 and good selectivity against memapsin 1. Inhibitor 5d has shown low nanomolar enzyme inhibitory potency (Ki=1.1 nM) and very good cellular inhibitory activity (IC50=39 nM). Furthermore, in a preliminary study, inhibitor 5d has shown 30% reduction of Abeta40 production in transgenic mice after a single intraperitoneal administration (8 mg/kg). A protein-ligand X-ray crystal structure of 5d-bound memapsin 2 provided vital molecular insight that can serve as an important guide to further design of novel inhibitors.