- Synthesis of 6- and 9-alkylaminomethyl furoflavones as gastroprotective agents.
Synthesis of 6- and 9-alkylaminomethyl furoflavones as gastroprotective agents.
European journal of medicinal chemistry (2007-03-10)
Fatma A Ragab, Ghaneya S Hassan, Hanan A Yossef, Hanna A Hashem
PMID17346862
ABSTRACT
The synthesis of 9- and 6-alkylaminomethyl furoflavones 5a, b, 9a-c, 13a, b, 15a-g and 18 from the naturally occurring chromones visnagin and khellin. Gastroprotective potency of these compounds in the ethanol damage model was determined. The results indicate that, through appropriate substitution, furoflavones can be obtained that are gastroprotective.