Merck
  • Conformational equilibria in formic acid and the adduct of formic acid and hexafluoroacetone, HCO2C(CF3)2OH.
Conformational equilibria in formic acid and the adduct of formic acid and hexafluoroacetone, HCO2C(CF3)2OH.
The Journal of organic chemistry (2007-02-17)
Diwakar M Pawar, Dalephine Cain-Davis, Eric A Noe
ABSTRACT
Low-temperature 1H and 13C NMR spectra of formic acid (1) showed separate signals for the E and Z conformations in solvents containing a hydrogen bond acceptor, dimethyl ether. The population of E-1 (6.2% in 3:1:1 CHClF2/CHCl2F/(CH3)2O) was larger than that for 13C-labeled methyl formate in the same solvent (0.2%), which indicated that the relative populations are not determined by steric effects. The free-energy difference between the E and Z conformations of 1 was 0.9 kcal/mol. In a 1:3 CD2Cl2/(CH3)2O solvent mixture, peaks for E and Z conformations were found at low temperatures by 1H and 13C NMR for both formic acid and an adduct with hexafluoroacetone, HCO2C(CF3)2OH (2). The population of E-1 in this solvent mixture was 4.3% by 13C NMR. The carbon spectrum showed two peaks in the carbonyl carbon region of nearly equal intensities at -151.6 degrees C, with E-2 (48%) absorbing downfield of the major Z-2 (52%). The large population of E-2 confirms that electron-withdrawing groups R' in RCO2R' enhance the populations of the E-isomers. The free-energy barriers for 2 of 6.24 (E-to-Z) and 6.26 kcal/mol (Z-to-E) were determined from rate constants obtained by line shape analysis at -143.2 degrees C.