Merck
Benzofused tricycles based on 2-quinoxalinol.
Journal of combinatorial chemistry (2007-01-09)
Gang Liu, Li Li, Binbin Kou, Suode Zhang, Liang Zhang, Yunyun Yuan, Tao Ma, Yan Shang, Yuancheng Li
ABSTRACT
This paper describes our recent efforts to synthesize novel compound scaffolds integrating 2-quinoxalinol with privileged structures of 1,3-dihydro-benzoimidazol-2-one, 1,3-dihydro-benzoimidazole-2-thione, 3-hydroxy-1H-quinoxalin-2-one, 2H-benzo[1,4]oxazin-3-ol, 2H-benzo[1,4]thiazin-3-ol, and 1,3,4,5-tetrahydro-benzo[1,4]diazepin-2-one, respectively. Eight novel benzofused tricycles and their substituent diversity points were developed. These include pyrazino[2,3-g]quinoxaline-2,8-diol (I), 3-hydroxy-6,8,9,10-tetrahydro-1,4,6,10-tetraaza-cyclohepta[b]naphthalen-7-one (II), 6-hydroxy-4H-1-oxa-4,5,8-triaza-anthracen-3-one (III), 6-hydroxy-4H-1-thia-4,5,8-triaza-anthracen-3-one (IV), 6-hydroxy-1,1-dioxo-1,4-dihydro-2H-1lambda(6)-thia-4,5,8-triaza-anthracen-3-one (V), 6-hydroxy-1,3-dihydro-imidazo[4,5-g]quinoxalin-2-one (VI), 6-hydroxy-1,3-dihydro-imidazo[4,5-g]quinoxaline-2-thione (VII), and 7-hydroxy-1,4-dihydro-pyrazino[2,3-g]quinoxaline-2,3-dione (VIII). This strategy of integrating two benzofused privileged structures into one molecule may provide a greater chance for the discovery of novel lead compounds.