Merck
  • RuCl3/PPh3: an efficient combination for the preparation of chiral 1,3-anti-diols through catalytic hydrogenation.
RuCl3/PPh3: an efficient combination for the preparation of chiral 1,3-anti-diols through catalytic hydrogenation.
Organic letters (2006-12-29)
Olivier Labeeuw, Christophe Roche, Phannarath Phansavath, Jean-Pierre Genêt
ABSTRACT
[reaction: see text] An efficient economical alternative to the commonly used Evans' reagent for the diastereoselective reduction of beta-hydroxy ketones is reported. Thus, ruthenium-mediated hydrogenation of enantioenriched beta-hydroxy ketones using RuCl3 associated to achiral monophosphines allowed the preparation of a series of 1,3-anti-diols in good yields and with a high level of diastereoselectivity. A short screening of ligands pointed out PPh3 as the most effective phosphine, and PCy3 afforded the 1,3-diols with an unexpected moderate syn selectivity.