Merck
  • Stability and plasticity of intrinsic membrane properties in hippocampal CA1 pyramidal neurons: effects of internal anions.
Stability and plasticity of intrinsic membrane properties in hippocampal CA1 pyramidal neurons: effects of internal anions.
The Journal of physiology (2006-12-02)
Catherine Cook Kaczorowski, John Disterhoft, Nelson Spruston
ABSTRACT
CA1 pyramidal neurons from animals that have acquired hippocampal tasks show increased neuronal excitability, as evidenced by a reduction in the postburst afterhyperpolarization (AHP). Studies of AHP plasticity require stable long-term recordings, which are affected by the intracellular solutions potassium methylsulphate (KMeth) or potassium gluconate (KGluc). Here we show immediate and gradual effects of these intracellular solutions on measurement of the AHP and basic membrane properties, and on the induction of AHP plasticity in CA1 pyramidal neurons from rat hippocampal slices. The AHP measured immediately after establishing whole-cell recordings was larger with KMeth than with KGluc. In general, the AHP in KMeth was comparable to the AHP measured in the perforated-patch configuration. However, KMeth induced time-dependent changes in the intrinsic membrane properties of CA1 pyramidal neurons. Specifically, input resistance progressively increased by 70% after 50 min; correspondingly, the current required to trigger an action potential and the fast afterdepolarization following action potentials gradually decreased by about 50%. Conversely, these measures were stable in KGluc. We also demonstrate that activity-dependent plasticity of the AHP occurs with physiologically relevant stimuli in KGluc. AHPs triggered with theta-burst firing every 30 s were progressively reduced, whereas AHPs elicited every 150 s were stable. Blockade of the apamin-sensitive AHP current (I(AHP)) was insufficient to block AHP plasticity, suggesting that plasticity is manifested through changes in the apamin-insensitive slow AHP current (sI(AHP)). These changes were observed in the presence of synaptic blockers, and therefore reflect changes in the intrinsic properties of the neurons. However, no AHP plasticity was observed using KMeth. In summary, these data show that KMeth produces time-dependent changes in basic membrane properties and prevents or obscures activity-dependent reduction of the AHP. In whole-cell recordings using KGluc, repetitive theta-burst firing induced AHP plasticity that mimics learning-related reduction in the AHP.