Merck
  • Spectrophotometric determination of trace arsenic in water samples using a nanoparticle of ethyl violet with a molybdate-iodine tetrachloride complex as a probe for molybdoarsenate.
Spectrophotometric determination of trace arsenic in water samples using a nanoparticle of ethyl violet with a molybdate-iodine tetrachloride complex as a probe for molybdoarsenate.
Analytical chemistry (2006-11-16)
Keisuke Morita, Emiko Kaneko
ABSTRACT
A new spectrophotometric method was developed for the determination of low ppb levels of arsenic in water. We found that Ethyl Violet with molybdate-iodine tetrachloride complex forms nanoparticles under acidic conditions, which provide a sensitive probe for molybdoarsenate. The nanoparticles form stable particles with a diameter micrometers in size in the presence of heteropolyacid, and the resulting particles give a purple color to the apparently homogeneous solution, the intensity of which depends on the arsenic concentration. The nanoparticle itself is unstable due to conversion of the dye to a colorless carbinol species under acidic conditions without heteropolyacid. Although triphenylmethane dyes have been the subject of a number of investigations, there do not appear to be any reports on the dye particles for trace determination. The calibration curve is linear up to 20 microg L-1 arsenic, and the detection limit is 0.5 microg L-1 (6.6 x 10(-9) mol L-1). The coefficient of variation for spectrophotometry at 10 microg L-1 is 5.8% (n = 8). Furthermore, it is possible to detect concentrations as low as 1 microg L-1 arsenic visually using this method. The interferences from phosphorus and silica were eliminated using an anion exchange column and sodium fluoride as a masking agent, respectively. The proposed method has been successfully applied to water samples in abandoned mine water, groundwater, and river water. There was good agreement between the results obtained by the proposed method and those by hydride generation atomic absorption spectrometry. Since this method is specific for As(V), it is applicable to the speciation of arsenic oxidation states. Our method has enormous practical potential for simple and field detection of arsenic, requiring no complex apparatus or skilled laboratory support.