Merck
  • A stereoselective synthesis of digitoxin and digitoxigen mono- and bisdigitoxoside from digitoxigenin via a palladium-catalyzed glycosylation.
A stereoselective synthesis of digitoxin and digitoxigen mono- and bisdigitoxoside from digitoxigenin via a palladium-catalyzed glycosylation.
Organic letters (2006-09-08)
Maoquan Zhou, George A O'Doherty
ABSTRACT
A convergent and stereocontrolled route to trisaccharide natural product digitoxin has been developed. The route is amenable to the preparation of both the digitoxigen mono- and bisdigitoxoside. This route featured the iterative application of the palladium-catalyzed glycosylation reaction, reductive 1,3-transposition, diastereoselective dihydroxylation, and regioselective protection. The natural product digitoxin was fashioned in 15 steps starting from digitoxigenin 2 and pyranone 8a or 18 steps from achiral acylfuran.