Merck
  • Structural studies on yeast 3-phosphoglycerate kinase. Identification by immuno-affinity chromatography of one glutamyl residue essential for yeast 3-phosphoglycerate kinase activity. Its location in the primary structure.
Structural studies on yeast 3-phosphoglycerate kinase. Identification by immuno-affinity chromatography of one glutamyl residue essential for yeast 3-phosphoglycerate kinase activity. Its location in the primary structure.
European journal of biochemistry (1980-04-01)
G Desvages, C Roustan, A Fattoum, L A Pradel
ABSTRACT
3-Phosphoglycerate kinase is inactivated by 1-cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate and nitrotyrosine ethyl ester. The coupling of 1 mol nitrotyrosine/mol enzyme is sufficient to inactivate the protein completely. A weak protection against inactivation is observed with each substrate added separately. In contrast, the complex ATP--3-phosphoglycerate--enzyme or ATP--Mg--3-phosphoglycerate--enzyme affords a considerable protection. The critical residue is identified as a glutamyl residue after isolation by immuno-affinity chromatography of nitrotyrosyl peptide resulting from exhaustive proteolytic digestion of the modified protein. In addition, the determination of the primary sequence of the C-terminal part of the protein leads to the location of the glutamyl residue at position eight from the C-terminus. We conclude that this glutamyl residue is situated in the domain which does not bind the nucleotide substrates [Bryant, T.N., Watson, H.C. and Wendell, P.L. (1974) Nature (Lond.) 247, 14--17]. Its role in the catalysis process is discussed.