- Biomolecular sensor based on fluorescence-labeled aptamer.
Biomolecular sensor based on fluorescence-labeled aptamer.
Bioorganic & medicinal chemistry letters (2006-06-08)
Hiroaki Ozaki, Akifumi Nishihira, Masayuki Wakabayashi, Masayasu Kuwahara, Hiroaki Sawai
PMID16757168
ABSTRACT
Fluorescent DNA probes for L-argininamide were developed by a combination of DNA aptamers and fluorophore-quencher pairs. These molecules were synthesized by a combination of pre- and post-synthetic modification methods. The fluorescence-labeled aptamer could detect L-argininamide specifically. The binding affinities were defined by the binding affinity of the original aptamer to indicate that the end labeling of the aptamer did not influence the affinities.