Merck
  • Analysis of chemical warfare agents III. Use of bis-nucleophiles in the trace level determination of phosgene and perfluoroisobutylene.
Analysis of chemical warfare agents III. Use of bis-nucleophiles in the trace level determination of phosgene and perfluoroisobutylene.
Journal of chromatography. A (2005-11-30)
Bob Muir, David B Cooper, Wendy A Carrick, Christopher M Timperley, Ben J Slater, Suzanne Quick
ABSTRACT
The reactivity of phosgene and perfluoroisobutylene (PFIB) towards 1,2-bis-nucleophiles was exploited to allow determination of these gases in air samples. 2-Aminothiophenol (ATP), 3,4-dimercaptotoluene (DMT) and 2-hydroxymethylpiperidine (HMP) were evaluated as bis-nucleophiles capable of forming thermally-stable derivatives with phosgene and PFIB when loaded with triethylamine onto Tenax TA. Experimental design was used to optimise thermal desorption conditions. Detection limits in the low ngm(-3) range were observed for the five derivatives investigated. This work represents the most sensitive analytical method for trace level quantitation of phosgene and PFIB published to date.
Sigma-Aldrich
2-Aminothiophenol, 99%