- Lipid-soluble 3-pyridinol antioxidants spare alpha-tocopherol and do not efficiently mediate peroxidation of cholesterol esters in human low-density lipoprotein.
Lipid-soluble 3-pyridinol antioxidants spare alpha-tocopherol and do not efficiently mediate peroxidation of cholesterol esters in human low-density lipoprotein.
Journal of medicinal chemistry (2005-10-28)
Hye-Young Kim, Derek A Pratt, Jennifer R Seal, Maikel Wijtmans, Ned A Porter
PMID16250637
ABSTRACT
Because alpha-tocopherol (alpha-TOH) mediates the peroxidation of cholesterol-esterified lipids in human low-density lipoprotein (LDL) in vitro and has displayed disappointing results against the onset and development of atherosclerosis, it may not be appropriate for use as a therapeutic in the prevention and/or treatment of the disease. Herein are described the experimental results demonstrating that 3-pyridinols spare alpha-TOH, do not efficiently mediate lipid peroxidation, and protect lipoprotein tryptophan residues in human LDL.