Merck
Photostability and photoprotection factor of boldine and glaucine.
Journal of photochemistry and photobiology. B, Biology (2005-06-21)
María Eliana Hidalgo, Miguel Farah, Leonardo Carrasco, Ernesto Fernández
ABSTRACT
Boldine hydrochloride was more photounstable than boldine after irradiation with UVB (lambda = 300 nm). However, photoconsumption quantum yields, for glaucine hydrochloride (6.5 x 10(-2)) and boldine hydrochloride (6.7 x 10(-2)) in air, were quite similar. The photolysis was oxygen dependent in both cases, and the effect over the kinetics after the addition of 2,2,6,6-tetramethyl-1-piperidinyloxy suggested free radicals participation. The fact that the antioxidative capacity of boldine and boldine hydrochloride did not change during the photolysis, suggests that the phenolic structure remains unchanged in the photoproducts, corroborated with the photoproducts analysis. The photoprotection capacity was evaluated before and after irradiation. Results indicate that the values before irradiation are similar for all three compounds, only glaucine increasing its capacity with length of irradiation time.