Merck
  • Adsorption of ethyl violet dye in aqueous solution by regenerated spent bleaching earth.
Adsorption of ethyl violet dye in aqueous solution by regenerated spent bleaching earth.
Journal of colloid and interface science (2005-06-01)
W T Tsai, Y M Chang, C W Lai, C C Lo
ABSTRACT
The adsorption of basic dye (i.e., ethyl violet or basic violet 4) from aqueous solution onto the regenerated spent bleaching earth (RSBE) was carried out by varying the process parameters such as initial concentration, pH, and temperature. As analytical comparisons, activated bleaching earth (ABE) was also used as adsorbent for the adsorption of the basic dye at various initial concentrations. The experimental results showed that the adsorption process can be well described with the pseudo-second-order reaction model and less fitted by the intra-particle diffusion model. The kinetic parameters of both models obtained in the present work are in line with pore properties of the two adsorbents. According to the equilibrium adsorption capacity from the fitting of pseudo-second-order reaction model, it was further found that the both models of Langmuir and Freundlich appeared to fit well the isotherm data. In addition, the thermodynamic parameters were evaluated based on the pseudo-second-order rate constants, showing that the adsorption of ethyl violet onto the RSBE is endothermic in nature.