- [A method for determining arsenic in biological material].
[A method for determining arsenic in biological material].
Sudebno-meditsinskaia ekspertiza (1992-01-01)
V P Poluianov
PMID1589891
ABSTRACT
Method of extraction-atomic absorption detection of arsenic in biologic material using naphthenic acid was developed and tested. Method can be used for arsenic detection in postmortem material. It differs from the known method by its accuracy, reliability and simple use.